Skip to Content
MilliporeSigma

C-059

Clozapine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H19ClN4
CAS Number:
Molecular Weight:
326.82
UNSPSC Code:
41116107
MDL number:
NACRES:
NA.24
EC Number:
200-659-6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CN1CCN(CC1)C2=Nc3cc(Cl)ccc3Nc4ccccc24

InChI

1S/C18H19ClN4/c1-22-8-10-23(11-9-22)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)21-18/h2-7,12,20H,8-11H2,1H3

InChI key

QZUDBNBUXVUHMW-UHFFFAOYSA-N

Gene Information

General description

A Certified Snap-N-Spike® Solution for use in clinical toxicology, forensic analysis, or urine drug testing applications by LC/MS or GC/MS. Clozapine, sold as Clozaril® and FazaClo® in the US, is an atypical antipsychotic drug used for treatment of schizophrenia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Clozaril is a registered trademark of Sandoz, Inc.
FazaClo is a registered trademark of Jazz Pharmaceuticals International III Limited
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Bianca A Silva et al.
Nature neuroscience, 16(12), 1731-1733 (2013-11-12)
The neural circuits mediating fear to naturalistic threats are poorly understood. We found that functionally independent populations of neurons in the ventromedial hypothalamus (VMH), a region that has been implicated in feeding, sex and aggression, are essential for predator and
Gary Remington et al.
Psychopharmacology, 225(3), 505-518 (2012-11-28)
Clozapine levels are advocated in the monitoring of patients on this drug and have now been used for a number of years. A safety-related threshold has also been proposed, as well as therapeutic lower and upper thresholds. While there has
Chun-Hsin Chen et al.
The Journal of clinical psychiatry, 74(5), e424-e430 (2013-06-14)
Many studies have shown that metformin can decrease body weight and improve metabolic abnormalities in patients with schizophrenia. Whether or not the beneficial effects can be sustained after discontinuation of metformin needs to be evaluated. We conducted a 24-week randomized



Global Trade Item Number

SKUGTIN
C-059-1ML04061833444603