Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z768278

Sigma-Aldrich

Aldrich® high-vacuum glass stockcock, T-bore

stopcock bore 2 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.79

material

glass plug

Joint

joint plug12/30

side arm size

9 mm

stopcock bore

2 mm

Looking for similar products? Visit Product Comparison Guide

General description

Stopcock has three side arms sealed 90° apart. The solid glass plug has a "T" shaped bore so that any two or all three side arms can be connected at one time. All sizes are supplied complete with a metal retaining device.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service