Key Documents
BR88665
BRAND® stopcock T-bore with nozzles
for tube I.D. 6 mm
Synonym(s):
PTFE, Stopcocks
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
material
PTFE
packaging
pack of 1 ea
manufacturer/tradename
BRAND 88665
I.D.
3 mm
General description
The BRAND® T-bore stopcock with nozzles is made from high-quality PTFE, providing outstanding chemical resistance for a wide range of laboratory applications. Featuring a valve lever constructed from polypropylene (PP), it ensures smooth operation and control. The stopcock is designed with integrated hose nozzles, facilitating efficient fluid transfer and easy connection to hoses. Its robust construction guarantees reliable performance, making it ideal for handling aggressive chemicals and solvents.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)