Documents
Z286478
Nalgene® stopcock
3-way, bore size 4 mm
Synonym(s):
3-way connector, hose connector
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
material
polypropylene (with TFE plug)
description
for tubing I.D. 1/4 -5/16 in.
feature
autoclavable
packaging
pack of 1 ea
case of 4 ea
manufacturer/tradename
Nalgene® 6470, 0004
bore size
4 mm
General description
Serrated tubulation on each end.
Features and Benefits
  • Autoclave friendly
  • Non-seizing, leakproof PTFE plug
  • Tubulated outlets are arranged in a T shape and accepts tubing of 1/4 in. 7/16 in.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries