Key Documents
Z341924
IKA® dish attachment for HS/KS 260 basic and control shakers
AS 260.3 dish attachment, 1/cs
Synonym(s):
benchtop shakers, lab shakers, laboratory dish shakers, laboratory shakers
About This Item
UNSPSC Code:
41121500
NACRES:
NB.73
material
foil (slip-resistant)
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 3120600
suitability
suitable for
General description
Note: For smooth movement for cell cultures, nutrient media in Petri dishes, culture bottles and vessels with a low center of gravity; with integrated slip-resistant foil (PP).
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)