Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z264407

Supelco

Hamilton® TLC syringes

1705, volume 50 μL, needle size 22 ga, needle L 51 mm (2 in.),

coated tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

material

stainless steel needle (PTFE coated tip)

needle size

22 ga

feature

needle type fixed

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 80950

needle L

51 mm (2 in.)

volume

50 μL

Looking for similar products? Visit Product Comparison Guide

General description

PTFE coats the final 3/4 in. of this 2 in. (51 mm) long cemented needle. This allows for more reproducible sample spotting on TLC plates.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service