Key Documents
HAM7653-01
Hamilton® TLC syringe
model, 1701 RN, no needle, volume 10 μL
About This Item
UNSPSC Code:
42142600
NACRES:
NB.03
material
Gastight syringe
feature
autoclavable
model 1701 RN, no needle
manufacturer/tradename
Hamilton
parameter
10-115 °C temp. range (50-239 °F)
1000 psig pressure
I.D. × O.D.
0.460 mm × 6.60 mm
volume
10 μL
Related Categories
General description
The needles on Thin Layer Chromatography (TLC) syringes are coated from the point for 3/4” with a PTFE-coating. The coating reduces the surface tension between the needle and the liquid making it ideal for reproducible sample spotting. The TLC syringe is available as a cemented needle (SNTLC) or as a replacement needle to be installed into a standard removable needle (RN) syringe. To find the appropriate no needle syringe select general syringes and filter by termination and gauge. Choose the “RN” and “No Needle Included” options to see all syringes that can receive TLC needles. To find the appropriate TLC needle select the TLC option under “Needles” in the main product menu.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)