Key Documents
Z219428
Hirsch filter funnel
porosity 25-50 μm (C)
About This Item
UNSPSC Code:
41104900
NACRES:
NB.43
manufacturer/tradename
Ace Glass 972703
disc diam.
10 mm
pore size
25-50 μm porosity (C)
capacity
7 mL capacity
General description
For use with all microscale flasks and vials with ST/NS: 14/10 joint. Plain stem funnel has a 10 mm diameter sintered disc with a capacity above disc of approximately 7 mL.
Choose from one of the most recent versions:
Certificates of Analysis (COA)