Key Documents
Z214558
Micro photochemical reaction assembly
with quartz well
About This Item
UNSPSC Code:
41121800
feature
CE compliant: no
CSA approved: no
manufacturer/tradename
Ace Glass 788060
availability
not available in EU, Canada
General description
New reactor design1 handles volumes from 5 to 15 mL. Borosilicate glass body with #25 Ace-Thred at top, 14/10 outer side joint externally threaded to allow an O-ring cap-seal connection with mating male joints, and a porosity E (4 to 8 μm) filter disc for use as a gas inlet to remove oxygen from the solution or for agitation. Includes an 11 mm I.D. jacketed quartz immersion well, and a quartz Pen-Ray 5.5 W, low pressure, cold cathode, mercury lamp with matching 115 V, 60 Hz power supply.
1.Designed by John H. Penn and Richard D. Orr, West Virginia Univ. Dept. of Chemistry, Morgantown, WV 26506.
Choose from one of the most recent versions:
Certificates of Analysis (COA)