Key Documents
SYNV190390
Synthware Double-Walled Quartz Photochemical Immersion Well,
L 390 mm
Synonym(s):
Synthware V190390, immersion well
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
packaging
pkg of 1 ea
manufacturer/tradename
Synthware V190390
L
390 mm
General description
Double-walled quartz immersion well for use with V150500 reactor and lamp. A small diameter inner tube extends down the annular space to insure flow of coolant from bottom of well upward to outlet. Well is used with RC50 Nylon Bushing with Viton® O-ring to form a compression type seal with threaded vessels. Well offers a greaseless connection and can be adjusted to desired height .The lack of the ground joint means easier dismantling since there is no freezing.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)