Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z136786

Flex-Grip® stirrer chuck

fits shaft o.d., 10 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.72

feature

fits shaft o.d. 10 mm

manufacturer/tradename

Ace Glass 812410

Looking for similar products? Visit Product Comparison Guide

General description

Nylon chuck for use with stirring shafts. Has flexible insert that allows for misalignment of shaft without danger of breakage. Fits 8mm motor shaft.

Legal Information

Flex-Grip is a registered trademark of Ace Glass, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service