Key Documents
Z268747
Scienceware® Safe-Lab PTFE stirrer bearing
joint: ST/NS 24/40
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
material
Viton® O-ring
Joint
joint: ST/NS24/40
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F20320-1661
General description
Stopper-Loc® nut allows stirrer bearing to be locked to outer ground-glass joint. Complete with Stopper-Loc nut and two PTFE inserts. For 10mm diam. polished glass or PTFE stirring shafts.
Legal Information
Safe-Lab is a trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Stopper-Loc is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)