Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

W488401

Sigma-Aldrich

6-Methyl-5-hepten-2-ol

greener alternative

natural, ≥95%, FG

Synonym(s):

Coriander heptenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
128.21
FEMA Number:
4884
Beilstein:
1720072
MDL number:
UNSPSC Code:
12164502
Flavis number:
02.124
NACRES:
NA.21
Organoleptic:
coriander; green; sweet
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
botanical
Agency:
follows IFRA guidelines
food allergen:
no known allergens

biological source

botanical

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.448 (lit.)

bp

78 °C/14 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

coriander; green; sweet

SMILES string

CC(O)CC\C=C(\C)C

InChI

1S/C8H16O/c1-7(2)5-4-6-8(3)9/h5,8-9H,4,6H2,1-3H3

InChI key

OHEFFKYYKJVVOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service