Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M48805

Sigma-Aldrich

6-Methyl-5-hepten-2-one

99%

Synonym(s):

2-Methyl-2-hepten-6-one, 2-Oxo-6-methylhept-5-ene, 6-Methyl-5-hepten-2-ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2COCH3
CAS Number:
Molecular Weight:
126.20
Beilstein:
1741705
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.439 (lit.)

bp

73 °C/18 mmHg (lit.)

density

0.858 g/mL at 20 °C
0.855 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=C\CCC(C)=O

InChI

1S/C8H14O/c1-7(2)5-4-6-8(3)9/h5H,4,6H2,1-3H3

InChI key

UHEPJGULSIKKTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olabimpe Y Olaide et al.
PLoS neglected tropical diseases, 13(6), e0007460-e0007460 (2019-06-11)
African trypanosomosis, primarily transmitted by tsetse flies, remains a serious public health and economic challenge in sub-Saharan Africa. Interventions employing natural repellents from non-preferred hosts of tsetse flies represent a promising management approach. Although zebras have been identified as non-preferred
Arão Cardoso Viana et al.
Chemistry Central journal, 12(1), 54-54 (2018-05-12)
A fast method for the identification and stability evaluation of the aggregation pheromone rhynchophorol, which is the main substance used for chemical communication by the beetle Rhynchophorus palmarum L., was validated. In addition, the technique was applied to the evaluation
Edna Pesis et al.
Journal of the science of food and agriculture, 90(12), 2114-2123 (2010-07-08)
Californian Granny Smith apples are very susceptible to bitter pit (BP) and superficial scald symptoms that develop during cold storage. The main preventive means are diphenylamine dipping and/or gaseous application of the ethylene inhibitor 1-methylcylclopropene (1-MCP), which is effective against
Inmaculada Romero et al.
Journal of agricultural and food chemistry, 65(21), 4314-4320 (2017-05-11)
Freeze injuries in olives are responsible for the 'frostbitten olives' sensory defect that is sometimes detected in virgin olive oil. This defect is becoming one of the most common negative attributes today because climate change has modified weather patterns. The
B D Whitaker et al.
Journal of agricultural and food chemistry, 48(6), 2040-2043 (2000-07-11)
Conjugated triene (CT) oxidation products of alpha-farnesene have long been thought to be involved in development of superficial scald in apple fruit. Early studies found that CT hydroperoxides and the volatile 6-methyl-5-hepten-2-one (MHO) are major in vitro autoxidation products of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service