Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W272507

Sigma-Aldrich

Methyl trans-2-nonenoate

≥97%, FG

Synonym(s):

Methyl 2-nonenoate, Methyl (E)-2-nonenoate, melon nonenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CHCO2CH3
CAS Number:
Molecular Weight:
170.25
FEMA Number:
2725
EC Number:
Council of Europe no.:
2099
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.234
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

description

Reg. Compliance: FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.444 (lit.)

bp

115 °C/21 mmHg (lit.)

density

0.895 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; waxy; fruity; sweet

SMILES string

[H]\C(CCCCCC)=C(\[H])C(=O)OC

InChI

1S/C10H18O2/c1-3-4-5-6-7-8-9-10(11)12-2/h8-9H,3-7H2,1-2H3/b9-8+

InChI key

ZWNPUELCBZVMDA-CMDGGOBGSA-N

Related Categories

General description

Methyl trans-2-nonenoate is one of the volatile compounds identified in mango wine.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of volatile compounds of mango wine.
Pino JA & Queris O.
Food Chemistry, 125(4), 1141-1146 (2011)
Gilles Brackman et al.
PloS one, 6(1), e16084-e16084 (2011-01-21)
Many bacteria, including Vibrio spp., regulate virulence gene expression in a cell-density dependent way through a communication process termed quorum sensing (QS). Hence, interfering with QS could be a valuable novel antipathogenic strategy. Cinnamaldehyde has previously been shown to inhibit

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service