Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W467314

Sigma-Aldrich

δ-Hexadecalactone

≥98%, FG

Synonym(s):

δ-Juniper lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H30O2
CAS Number:
Molecular Weight:
254.41
FEMA Number:
4673
MDL number:
UNSPSC Code:
12164502
Flavis number:
10.049
NACRES:
NA.21

grade

FG
Halal
Kosher

Quality Level

reg. compliance

EU 1334/2008 & 178/2002
FDA 21 CFR 117

description

Organoleptic:
Class: Fatty
SubClass: Other

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

butter; waxy

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service