Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W468501

Sigma-Aldrich

δ-Tridecalactone

≥95%, FG

Synonym(s):

Tetrahydro-6-octyl-2H-pyran-2-one, 6-octyloxan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24O2
CAS Number:
Molecular Weight:
212.33
FEMA Number:
4685
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.058
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥95%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; dairy; milk; musty; buttery

SMILES string

O1C(CCCC1=O)CCCCCCCC

InChI

1S/C13H24O2/c1-2-3-4-5-6-7-9-12-10-8-11-13(14)15-12/h12H,2-11H2,1H3

InChI key

RZZLMGATMUAJPX-UHFFFAOYSA-N

Related Categories

General description

δ-Tridecalactone participates as an internal standard in a study on the conversion of linoleic acid to hydroxyl fatty acid by lactic acid bacterial strains. Its racemic form has been used in the synthesis of racemic-N-alkyl-5-acetoxyalkanamides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

259.7 °F

Flash Point(C)

126.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasutaka Shimotori et al.
Journal of oleo science, 64(11), 1213-1226 (2015-10-16)
A combination of Novozym 435-catalyzed methanolysis and amidation using racemic N-methyl-5-acetoxytridecan- and tetradecanamides as a substrate proceeded in good enantioselectivity to afford the corresponding (R)-N-methyl-5-acetoxyalkanamides, (S)-N-methyl-5-hydroxyalkanamides, and (S)-N-cyclohexyl-5-hydroxyalkanamides. Both enantiomers of δ-tri- and δ-tetradecalactones were synthesized in over 90% enantiomeric
Noriaki Kishimoto et al.
Lipids, 38(12), 1269-1274 (2004-02-12)
Twenty-three of 86 strains of lactic acid bacteria transformed linoleic acid into hydroxy FA. Two distinct conversion pathways were in operation. Two strains of Lactobacillus acidophilus and a strain of Pediococcus pentosaceus produced 13(S)-hydroxy-9-octadecenoic acid 113(S)-OH 18:11 and 10,13dihydroxyoctadecanoic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service