Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W435600

Sigma-Aldrich

trans-3-Hexen-1-ol

≥95%, stabilized

Synonym(s):

trans-3-Hexenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CH2OH
CAS Number:
Molecular Weight:
100.16
FEMA Number:
4356
Beilstein:
1719713
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009

Assay

≥95%

contains

alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.439 (lit.)

bp

61-62 °C/12 mmHg (lit.)

density

0.817 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green

SMILES string

[H]\C(CC)=C(\[H])CCO

InChI

1S/C6H12O/c1-2-3-4-5-6-7/h3-4,7H,2,5-6H2,1H3/b4-3+

InChI key

UFLHIIWVXFIJGU-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigehiro Namiki et al.
Journal of comparative physiology. A, Neuroethology, sensory, neural, and behavioral physiology, 194(5), 501-515 (2008-04-05)
Pheromone-source orientation behavior can be modified by coexisting plant volatiles. Some host plant volatiles enhance the pheromonal responses of olfactory receptor neurons and increase the sensitivity of orientation behavior in the Lepidoptera species. Although many electrophysiological studies have focused on
Damon J Crook et al.
Journal of economic entomology, 105(2), 429-437 (2012-05-23)
Field trapping assays were conducted in 2009 and 2010 throughout western Michigan, to evaluate lures for adult emerald ash borer, A. planipennis Fairmaire (Coleoptera: Buprestidae). Several ash tree volatiles were tested on purple prism traps in 2009, and a dark
Andre F Cruz et al.
Phytochemistry, 78, 72-80 (2012-04-24)
Fusarium diseases cause major economic losses in wheat-based crop rotations. Volatile organic compounds (VOC) in wheat and rotation crops, such as chickpea, may negatively impact pathogenic Fusarium. Using the headspace GC-MS method, 16 VOC were found in greenhouse-grown wheat leaves:
L Chen et al.
Bulletin of entomological research, 97(5), 515-522 (2007-10-06)
Parasitoids employ different types of host-related volatile signals for foraging and host-location. Host-related volatile signals can be plant-based, originate from the herbivore host or produced from an interaction between herbivores and their plant host. In order to investigate potential sex-
Sayaka Fujita et al.
Hormones and behavior, 58(2), 264-272 (2010-03-20)
Chronic maternal stress during pregnancy results in the "prenatally stressed" offspring displaying behavioral and neuroendocrine alterations that persist into adulthood. We investigated how inhalation of green odor (a mixture of equal amounts of trans-2-hexenal and cis-3-hexenol) by stressed dams might

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service