Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

W318844

Sigma-Aldrich

2-Methyl-3-furanthiol

natural, 1% in Sunflower oil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
FEMA Number:
3188
MDL number:
UNSPSC Code:
12164502
Flavis number:
13.055
NACRES:
NA.21

biological source

botanical

grade

Halal
Kosher
natural

vapor density

>1 (vs air)

refractive index

n20/D 1.518 (lit.)

bp

57-60 °C/44 mmHg (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; roasted; sulfurous

storage temp.

2-8°C

SMILES string

Cc1occc1S

InChI

1S/C5H6OS/c1-4-5(7)2-3-6-4/h2-3,7H,1H3

InChI key

RUYNUXHHUVUINQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service