Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W202800

Sigma-Aldrich

Allyl disulfide

≥80%, FG

Synonym(s):

Diallyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2SSCH2CH=CH2
CAS Number:
Molecular Weight:
146.27
FEMA Number:
2028
Beilstein:
1699241
EC Number:
Council of Europe no.:
485
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.008
NACRES:
NA.21
Organoleptic:
garlic; onion; alliaceous
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

>5 (vs air)

vapor pressure

1 mmHg ( 20 °C)

Assay

≥80%

refractive index

n20/D 1.541 (lit.)

bp

180-195 °C (lit.)

density

1.008 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

garlic; onion; alliaceous

SMILES string

C=CCSSCC=C

InChI

1S/C6H10S2/c1-3-5-7-8-6-4-2/h3-4H,1-2,5-6H2

InChI key

PFRGXCVKLLPLIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Allyl disulfide is naturally found in Allium species such as garlic and onion.[1] It is used as a flavoring agent in meat and condiments.[2]

Application


  • Paving New Roads Using Allium sativum as a Repurposed Drug and Analyzing its Antiviral Action Using Artificial Intelligence Technology.: Investigates the antiviral properties of Allium sativum (garlic), which is known to contain allyl disulfide. This study utilizes AI technology to analyze the mechanisms and efficacy of garlic as a repurposed drug for treating viral infections (Atoum MF et al., 2022).

Biochem/physiol Actions

Organosulfur compound that is naturally found in garlic. Contributes to the pungent odor of crushed garlic. Anticancer agent and may provide protection against cardiovascular disease.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fenaroli G
Fenaroli's Handbook of Flavor Ingredients, 17-17 (1971)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 87-87 (1997)
Nathaniel Edward Bennett Saidu et al.
Bioorganic & medicinal chemistry, 20(4), 1584-1593 (2012-01-24)
Coumarins and coumarin derivatives as well as diallyl polysulfides are well known as anticancer drugs. In order to find new drugs with anticancer activities, we combined coumarins with polysulfides in the form of di-coumarin polysulfides. These novel compounds were tested
R Arunkumar et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(10), 912-923 (2012-06-29)
Diallyl Disulfide (DADS) is one of the major components of garlic, which inhibits the proliferation of various cancer cells. Our previous studies showed that DADS inhibits cell growth and induces apoptosis on prostate cancer cells. Insulin like growth factor signaling
Hye Young Park et al.
Toxicology and applied pharmacology, 262(2), 177-184 (2012-05-09)
Diallyl disulfide (DADS), a main organosulfur component responsible for the diverse biological effects of garlic, displays a wide variety of internal biological activities. However, the cellular and molecular mechanisms underlying DADS' anti-inflammatory activity remain poorly understood. In this study, therefore

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service