Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W401755

Sigma-Aldrich

Bacon dithiazine

solution in EtOH

Synonym(s):

2,4,6-Tris(2-methylpropyl)-1,3,5-dithiazinane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H31NS2
CAS Number:
Molecular Weight:
289.54
FEMA Number:
4017
UNSPSC Code:
12164502
Flavis number:
15.113
NACRES:
NA.21
Organoleptic:
bacon; meaty; smoky
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

Kosher

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

bacon; meaty; smoky

SMILES string

S1C(SC(NC1CC(C)C)CC(C)C)CC(C)C

InChI

1S/C15H31NS2/c1-10(2)7-13-16-14(8-11(3)4)18-15(17-13)9-12(5)6/h10-16H,7-9H2,1-6H3

InChI key

RQGPQWUKHADVPF-UHFFFAOYSA-N

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service