Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W378704

Sigma-Aldrich

2-Methyl-3-tetrahydrofuranthiol

mixture of cis and trans, ≥97%, FG

Synonym(s):

2-methyloxolane-3-thiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10OS
CAS Number:
Molecular Weight:
118.20
FEMA Number:
3787
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
13.160
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

≥97%

refractive index

n20/D 1.488 (lit.)

bp

160-168 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

meaty; sulfurous

SMILES string

CC1OCCC1S

InChI

1S/C5H10OS/c1-4-5(7)2-3-6-4/h4-5,7H,2-3H2,1H3

InChI key

DBPHPBLAKVZXOY-UHFFFAOYSA-N

Related Categories

General description

2-Methyl-3-tetrahydrofuranthiol is a sulfur-containing flavor compound mainly used in meat flavorings.
Natural occurrence: Beef.

Biochem/physiol Actions

Odor at 0.1% PG
Taste at 0.1-0.5 ppm

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfur-containing furans in commercial meat flavorings.
Ruther J & Baltes W.
Journal of Agricultural and Food Chemistry, 42(10), 2254-2259 (1994)
Mosciano, G.
Perfumer & Flavorist, 26, 68-68 (2001)
Yuping Liu et al.
Molecules (Basel, Switzerland), 15(8), 5104-5111 (2010-08-18)
Five sulfur-containing flavor compounds were synthesized for the first time by the reaction of 4-ethyloctanoyl chloride with sulfur-containing alcohols or mercaptans. The synthesized compounds are 3-(methylthio)propyl 4-ethyloctanoate, 2-methyl-3-tetrahydro-furanthiol 4-ethyloctanoate, 4-methyl-5-thiazoleethanol 4-ethyloctanoate, 2-furan-methanethiol 4-ethyloctanoate and 2-methyl-3-furanthiol 4-ethyloctanoate. These five synthetic sulfur-containing
Sílvia Petronilho et al.
Molecules (Basel, Switzerland), 25(2) (2020-01-16)
Wine aroma is the result of complex interactions between volatile compounds and non-volatile ones and individual perception phenomenon. In this work, an aroma network approach, that links volatile composition (chromatographic data) with its corresponding aroma descriptors was used to explain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service