Skip to Content
MilliporeSigma
All Photos(2)

Documents

W474815

Sigma-Aldrich

2,4,6-triethyl-1,3,5-dithiazinane

≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NS2
CAS Number:
Molecular Weight:
205.38
FEMA Number:
4748
UNSPSC Code:
12164502
Flavis number:
15.054
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥95%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; meaty; brown; roasted; sulfurous

InChI

1S/C9H19NS2/c1-4-7-10-8(5-2)12-9(6-3)11-7/h7-10H,4-6H2,1-3H3

InChI key

AANIECMHWBXFCP-UHFFFAOYSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service