Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W368903

Sigma-Aldrich

cis-3-Hexenyl cis-3-hexenoate

≥95%, stabilized, FG

Synonym(s):

(Z)-3-hexen-1-yl (Z)-3-hexenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH=CHCH2CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
196.29
FEMA Number:
3689
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.291
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

Assay

≥95%

contains

alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.452 (lit.)

bp

60 °C/0.5 mmHg (lit.)

density

0.907 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

fresh; green; fruity; ethereal; pear

SMILES string

[H]\C(CC)=C(/[H])CCOC(=O)C\C([H])=C(\[H])CC

InChI

1S/C12H20O2/c1-3-5-7-9-11-14-12(13)10-8-6-4-2/h5-8H,3-4,9-11H2,1-2H3/b7-5-,8-6-

InChI key

UZJQQWFHPLYECS-SFECMWDFSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

210.0 °F - closed cup

Flash Point(C)

98.90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service