Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W340399

Sigma-Aldrich

cis-3-Hexenyl hexanoate

natural, ≥95%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
198.30
FEMA Number:
3403
MDL number:
UNSPSC Code:
12164502
Flavis number:
9.271
NACRES:
NA.21
Organoleptic:
green; waxy; fruity; tropical
grade:
FG
Fragrance grade
Halal
Kosher
natural
Agency:
follows IFRA guidelines
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

refractive index

n20/D 1.473 (lit.)

bp

115 °C/15 mmHg (lit.)

density

0.88 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; waxy; fruity; tropical

SMILES string

[H]\C(CC)=C(/[H])CCOC(=O)CCCCC

InChI

1S/C12H22O2/c1-3-5-7-9-11-14-12(13)10-8-6-4-2/h5,7H,3-4,6,8-11H2,1-2H3/b7-5-

InChI key

RGACQXBDYBCJCY-ALCCZGGFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service