Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W239402

Sigma-Aldrich

α,α-Dimethylphenethyl butyrate

≥95%, FCC, FG

Synonym(s):

(1-cyclohexyl-2-methylpropan-2-yl) butanoate, Benzyl dimethyl carbinol butyrate, Benzyl dimethyl carbinyl butyrate, Dimethyl benzyl carbinyl butyrate, dmbc butyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2C(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
220.31
FEMA Number:
2394
EC Number:
Council of Europe no.:
2084
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.232
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets requirements for JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥95%

refractive index

n20/D 1.4839 (lit.)

bp

237-255 °C (lit.)

density

0.969 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity; floral; herbaceous

SMILES string

CCCC(=O)OC(C)(C)Cc1ccccc1

InChI

1S/C14H20O2/c1-4-8-13(15)16-14(2,3)11-12-9-6-5-7-10-12/h5-7,9-10H,4,8,11H2,1-3H3

InChI key

SHSGYHAHMQLYRB-UHFFFAOYSA-N

Related Categories

General description

α,α-Dimethylphenethyl butyrate belongs to the fragrance structural group aryl alkyl alcohol simple acid esters (AAASAE). The compound has a mild, herbaceous odor. It is a fragrance ingredient used in various non-cosmetic products, such as detergents, household cleansers, and shampoos. It is also found in decorative cosmetics, toiletries, and shampoos.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

242.6 °F - closed cup

Flash Point(C)

117 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragrance material review on 1, 1-dimethyl-2-phenylethyl butyrate
McGinty D, et al.
Food And Chemical Toxicology, 50, S439-S442 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service