Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W251801

Sigma-Aldrich

Geranyl isovalerate

≥95%, stabilized, FG

Synonym(s):

Geranyl 3-methyl butanoate, Trans-3,7-dimethyl-2,6-octadien-1-yl isovalerate, [(3E)-3,7-dimethylocta-3,6-dienyl] 3-methylbutanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
238.37
FEMA Number:
2518
EC Number:
Council of Europe no.:
448
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.453
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥95%

contains

alpha-tocopherol, synthetic as stabilizer

composition

contains IFRA and EU 1223/2009 restricted Geraniol

refractive index

n20/D 1.458 (lit.)

bp

279 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

geraniol

Organoleptic

apple; blueberry; green; fruity; pineapple

SMILES string

CC(C)CC(=O)OC\C=C(/C)CC\C=C(\C)C

InChI

1S/C15H26O2/c1-12(2)7-6-8-14(5)9-10-17-15(16)11-13(3)4/h7,9,13H,6,8,10-11H2,1-5H3/b14-9+

InChI key

SOUKTGNMIRUIQN-NTEUORMPSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service