Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W244201

Sigma-Aldrich

Ethyl levulinate

≥98%, FCC, FG

Synonym(s):

Ethyl 4-oxopentanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2COOC2H5
CAS Number:
Molecular Weight:
144.17
FEMA Number:
2442
Beilstein:
507641
EC Number:
Council of Europe no.:
373
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.435
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.422 (lit.)

bp

93-94 °C/18 mmHg (lit.)

density

1.016 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; green; melon; fruity; sweet

SMILES string

CCOC(=O)CCC(C)=O

InChI

1S/C7H12O3/c1-3-10-7(9)5-4-6(2)8/h3-5H2,1-2H3

InChI key

GMEONFUTDYJSNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanjin Im et al.
Bioresource technology, 193, 386-392 (2015-07-06)
This work addresses an unprecedented way of co-producing biodiesel (FAEE) and valuable chemicals of ethyl levulinate (EL), ethyl formate (EF) and diethyl ether (DEE) from wet in situ transesterification of microalgae. EL, EF, and DEE were significantly produced up to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service