Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

W267215

Sigma-Aldrich

4-(4-Methoxyphenyl)-2-butanone

natural, ≥96%, FG

Synonym(s):

Anisylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CH2COCH3
CAS Number:
Molecular Weight:
178.23
FEMA Number:
2672
Beilstein:
1869592
MDL number:
UNSPSC Code:
12164502
Flavis number:
7.029
NACRES:
NA.21
grade:
FG
Kosher
natural
biological source:
botanical
food allergen:
no known allergens

biological source

botanical

grade

FG
Kosher
natural

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥96%

refractive index

n20/D 1.519 (lit.)

bp

152-153 °C/15 mmHg (lit.)

mp

8 °C (lit.)

density

1.046 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; floral; raspberry; sweet

SMILES string

COc1ccc(CCC(C)=O)cc1

InChI

1S/C11H14O2/c1-9(12)3-4-10-5-7-11(13-2)8-6-10/h5-8H,3-4H2,1-2H3

InChI key

PCBSXBYCASFXTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service