Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W210218

Sigma-Aldrich

Anisyl propionate

≥98%, FG

Synonym(s):

4-Methoxybenzyl propionate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CO2CH2C6H4OCH3
CAS Number:
Molecular Weight:
194.23
FEMA Number:
2102
EC Number:
Council of Europe no.:
426
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.145
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.508 (lit.)

bp

277 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; floral; vanilla

SMILES string

CCC(=O)OCc1ccc(OC)cc1

InChI

1S/C11H14O3/c1-3-11(12)14-8-9-4-6-10(13-2)7-5-9/h4-7H,3,8H2,1-2H3

InChI key

YWIJRJQYADFRTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service