Key Documents
TEM-LC305CU
Sigma-Aldrich
Lacey Carbon Supported Copper Grids
hole size 50 nm, box of 5
Synonym(s):
LACEY CARBON FIL - CU 300 MESH 100 MICRON 5/BX
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 5
hole size
50 nm
General description
Lacey carbon film supported TEM grids. This carbon coating consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Choose from one of the most recent versions:
Certificates of Analysis (COA)