Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SYNF891425

Synthware reaction tube with glass stopcock

25 mL, joint: ST/NS 14/20

Synonym(s):

Synthware F891425

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121700
NACRES:
NB.51

material

colorless tube
glass tube

Joint

joint: ST/NS14/20

sterility

non-sterile

feature

graduations: no
Push fit

packaging

pack of 1 ea

manufacturer/tradename

Synthware F891425

capacity

25 mL
25 mL

Looking for similar products? Visit Product Comparison Guide

General description

With an outer joint and 2 mm glass stopcock having a constant pressure retention device.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service