Key Documents
TEM-FF100-NI-UL
Sigma-Aldrich
Ultral-Thin Formvar Supported Nickel Square Mesh
size 100 mesh, box of 50
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
packaging
box of 50
size
100 mesh
General description
A 5-6 nm ultral-thin formvar film supported standard square mesh grids. Nickel grid, 100 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)