Documents
BAF182370000
Dialysis bag holder clip for beakers
pack of 6 ea
About This Item
UNSPSC Code:
41123100
NACRES:
NB.51
material
polypropylene
feature
autoclavable
packaging
pack of 6 ea
manufacturer/tradename
Bel-Art F18237-0000
General description
The bag holder is resistant to chemicals and may be autoclaved.
  • Suspend dialysis/osmosis bag in fluid
  • Sturdy holder clips onto the side of an open container with a wall up to 1/4 in. thick
  • Holder has room for sample identification marks