Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T83607

Sigma-Aldrich

Bromotriphenylmethane

98%

Synonym(s):

Triphenylbromomethane, Trityl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3CBr
CAS Number:
Molecular Weight:
323.23
Beilstein:
1878494
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

bp

230 °C/15 mmHg (lit.)

mp

152-154 °C (lit.)

storage temp.

2-8°C

SMILES string

BrC(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C19H15Br/c20-19(16-10-4-1-5-11-16,17-12-6-2-7-13-17)18-14-8-3-9-15-18/h1-15H

InChI key

NZHXEWZGTQSYJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reagent for the preparation of trityl esters.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siddabasave Gowda B Gowda et al.
Lipids, 55(4), 395-401 (2020-05-15)
An efficient three-step strategy for the convenient synthesis of Sn-glycero-3-phosphoethanolamine (GroPEtn) from a commercially available 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine (DPPE) is reported. Direct hydrolysis of DPPE produces a complex inseparable mixture, hence a protection and deprotection strategy is employed to prepare GroPEtn. The
Cansu Aydogan et al.
Macromolecular rapid communications, 40(14), e1900109-e1900109 (2019-05-16)
A straightforward transformation protocol combining two distinct living polymerization methods for the controlled synthesis of block copolymers is described. In the first step, bromo-terminated poly(methyl methacrylate) is prepared by atom transfer radical polymerization (ATRP). Then, a bromide end group is
Yusuke Yamamoto et al.
Analytical and bioanalytical chemistry, 413(1), 245-254 (2020-10-23)
Lysophosphatidylethanolamines (LysoPEs) are the partial hydrolysis products of phosphatidylethanolamine. Despite the unique in vitro bioactivities of LysoPEs, there are limited reports on the pathophysiological role of LysoPEs in the serum, due to the lack of sensitive analytical methods for determination
Krzysztof Matyjaszewski et al.
Molecules (Basel, Switzerland), 26(2) (2021-01-14)
N-alkyl phosphoranimines were synthesized via the Staudinger reaction of four different alkyl azides with tris(2,2,2-trifluoroethyl) phosphite. N-adamantyl, N-benzyl, N-t-butyl, and N-trityl phosphoranimines were thoroughly characterized and evaluated as chain-capping compounds in the anionic polymerization of P-tris(2,2,2-trifluoroethoxy)-N-trimethylsilyl phosphoranimine monomer. All four
Salvatore Debonis et al.
Journal of medicinal chemistry, 51(5), 1115-1125 (2008-02-13)
The human kinesin Eg5 is a potential drug target for cancer chemotherapy. Eg5 specific inhibitors cause cells to block in mitosis with a characteristic monoastral spindle phenotype. Prolonged metaphase block eventually leads to apoptotic cell death. S-trityl-L-cysteine (STLC) is a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service