Skip to Content
MilliporeSigma
All Photos(1)

Documents

164577

Sigma-Aldrich

Tritylium tetrafluoroborate

Synonym(s):

Triphenylcarbenium tetrafluoroborate, Trityl fluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3C(BF4)
CAS Number:
Molecular Weight:
330.13
Beilstein:
3659536
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

205-215 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.c1ccc(cc1)[C+](c2ccccc2)c3ccccc3

InChI

1S/C19H15.BF4/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;2-1(3,4)5/h1-15H;/q+1;-1

InChI key

VQXBOEYKSVVPSP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Synthesis of substituted dihydroazulene photoswitches
  • Carbene-based Lewis pairs for hydrogen activation
  • 1,3-Dipolar cycloaddition reactions for preparation of α−amino-β-hydroxy esters
  • Oxidation of allenic compounds
  • Mukaiyama aldol addition reactions
  • Ionic hydrogenation as a counteranion and ligand source

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service