Skip to Content
MilliporeSigma
All Photos(1)

Documents

18735

Sigma-Aldrich

Hydrobromic acid solution

33 wt. % in acetic acid

Synonym(s):

Hydrogen bromide in acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HBr
Molecular Weight:
80.91
Beilstein:
3587158
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

Assay

31.5-34.5% (T)

form

liquid

concentration

33 wt. % in acetic acid

impurities

≤1% water (KFT)

density

1.40 g/mL at 20 °C

SMILES string

Br

InChI

1S/BrH/h1H

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrobromic acid solution (HBr) is a clear, yellow or brown colored liquid. Its reaction with K has been studied by a molecular beam technique.

Application

Hydrobromic acid solution may be used for the demethylation of 3,4-dimethoxy-6-bromonitrobenzene. Along with paraldehyde, it forms an efficient bromomethylating agent for benzene derivatives.
Hydrobromic acid solution may be used to infrared spectroscopic analysis of degree of N-deacetylation of chitosan.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Regioselective bromomethylation of 1, 2-dialkylbenzenes.
Garg N and Lee TR
Synlett, 9(03), 310-312 (1998)
Hydrogen Bromide-Acetic Acid Demethylation of 2, 3-Dimethoxy-6-bromobenzoic Acid. An Example of Concomitant Bromine Migration.
Pettit GR and Piatak DM.
The Journal of Organic Chemistry, 25(5), 721-725 (1960)
Study of chemical reaction mechanisms with molecular beams. The reaction of K with HBr.
Taylor EH and Datz S.
J. Chem. Phys. , 23(9), 1711-1718 (1955)
Improved infrared spectroscopic method for the analysis of degree of N-deacetylation of chitosan.
Sabnis S and Block LH.
Polym. Bull., 39(1), 67-71 (1997)
Yaohui Tang et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 34(7), 1138-1147 (2014-04-10)
Neural stem cells (NSCs) show therapeutic potential for ischemia in young-adult animals. However, the effect of aging on NSC therapy is largely unknown. In this work, NSCs were transplanted into aged (24-month-old) and young-adult (3-month-old) rats at 1 day after

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service