Skip to Content
MilliporeSigma
All Photos(2)

Documents

T27855

Sigma-Aldrich

2-Thiopheneethanol

98%

Synonym(s):

2-(2-Hydroxyethyl)thiophene, 2-(2-Thienyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8OS
CAS Number:
Molecular Weight:
128.19
Beilstein:
106985
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.551 (lit.)

bp

108-109 °C/13 mmHg (lit.)

density

1.153 g/mL at 25 °C (lit.)

SMILES string

OCCc1cccs1

InChI

1S/C6H8OS/c7-4-3-6-2-1-5-8-6/h1-2,5,7H,3-4H2

InChI key

VMJOFTHFJMLIKL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radoslaw Laufer et al.
Bioorganic & medicinal chemistry, 22(17), 4968-4997 (2014-07-22)
TTK kinase was identified by in-house siRNA screen and pursued as a tractable, novel target for cancer treatment. A screening campaign and systematic optimization, supported by computer modeling led to an indazole core with key sulfamoylphenyl and acetamido moieties at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service