Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

128643

Sigma-Aldrich

2-Pyridineethanol

98%

Synonym(s):

2-(2-Hydroxyethyl)pyridine, 2-(2-Pyridyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9NO
CAS Number:
Molecular Weight:
123.15
Beilstein:
111205
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.537 (lit.)

bp

114-116 °C/9 mmHg (lit.)

density

1.093 g/mL at 25 °C (lit.)

SMILES string

OCCc1ccccn1

InChI

1S/C7H9NO/c9-6-4-7-3-1-2-5-8-7/h1-3,5,9H,4,6H2

InChI key

BXGYBSJAZFGIPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(2-pyridyl)ethanol derivatives react with the aryl and alkenyl chlorides (palladium-catalyzed reactions) that result in the substitution of the chloro moieties with a 2-pyridylmethyl group.

Application

2-Pyridineethanol was used to prepare mononuclear nickel(II) acetate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalena Malik et al.
Journal of inorganic biochemistry, 215, 111311-111311 (2020-11-29)
Gold(III) complex containing 2-pyridineethanol has been synthesized and characterized structurally by single crystal X-ray diffraction, vibrational spectroscopy, 1H NMR spectroscopy, electrochemical study, and DFT calculations. The Au(III) ion is four coordinated with one N-donor ligand (L) and three Cl anions.
Miha Trdin et al.
Acta chimica Slovenica, 59(3), 478-483 (2013-09-26)
A new mononuclear nickel(II) acetate with 2-pyridineethanol, Ni(ac)2(2-PyEtOH)2 has been prepared. The reaction product is a mixture of two polymorphic forms that crystallize concomitantly: triclinic (PĪ) and monoclinic (P21/c). Their structures have been determined at 150 K. The molecular structure
Palladium-catalyzed 2-pyridylmethyl transfer from 2-(2-pyridyl)ethanol derivatives to organic halides by chelation-assisted cleavage of unstrained C(sp3 )--C(sp3) bonds.
Takashi Niwa et al.
Angewandte Chemie (International ed. in English), 46(15), 2643-2645 (2007-03-03)
Sara Ranjbar et al.
Toxicology and applied pharmacology, 362, 136-149 (2018-11-06)
Multidrug resistance (MDR) in cancer cells is often associated with overexpression of ATP-binding cassette (ABC) transporters, including P-glycoprotein (P-gp/ABCB1), multidrug resistance-associated protein 1 (MRP1/ABCC1) and breast cancer resistance protein (BCRP/ABCG2). Modulators of these transporters might be helpful in overcoming MDR.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service