Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

228796

Sigma-Aldrich

3-Thiopheneethanol

99%

Synonym(s):

2-(3-Thienyl)ethanol, 3-(2-Hydroxyethyl)thiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8OS
CAS Number:
Molecular Weight:
128.19
Beilstein:
107080
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.552 (lit.)

bp

110-111 °C/14 mmHg (lit.)

density

1.144 g/mL at 25 °C (lit.)

SMILES string

OCCc1ccsc1

InChI

1S/C6H8OS/c7-3-1-6-2-4-8-5-6/h2,4-5,7H,1,3H2

InChI key

YYPNNBPPDFTQFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Thiopheneethanol [3-(2-hydroxyethyl)thiophene] was used in the synthesis of various ether and ester derivatives such as 3-(2-(benzyloxy)ethyl)thiophene, 3-[2-((triphenylmethyl)oxy)ethyl]thiophene, 3-[2-((trimethylsilyl)oxy)ethyl]thiophene, 3-[2-((dimethyl-tert-butylsilyl)oxy)ethyl]thiophene, 3-(2-acetoxyethyl)thiophene and 3-(2-(benzoyloxy)ethyl)thiophene.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactive groups on polymer-covered electrodes. 5. Synthesis and cyclovoltammetric analysis of 3-substituted thiophene derivatives.
Macromolecules, 30(24), 7419-7426 (1997)
Consuelo Ripoll et al.
Pharmaceutics, 13(2) (2021-03-07)
Recently, it was proposed that the thiophene ring is capable of promoting mitochondrial accumulation when linked to fluorescent markers. As a noncharged group, thiophene presents several advantages from a synthetic point of view, making it easier to incorporate such a
Yuwei Hao et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 19(16), 2046-2051 (2018-03-25)
Highly efficient cell capture and release with low background are urgently required for early diagnosis of diseases such as cancer. Herein, we report an electrochemical responsive superhydrophilic surface exhibiting specific cell capture and release with high yields and extremely low

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service