Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

T26727

Sigma-Aldrich

Tetraphenyltin

97%

Synonym(s):

Tin tetraphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sn(C6H5)4
CAS Number:
Molecular Weight:
427.13
Beilstein:
3145842
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

14.7 (vs air)

Assay

97%

bp

>420 °C (lit.)

mp

224-227 °C (lit.)

SMILES string

c1ccc(cc1)[Sn](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/4C6H5.Sn/c4*1-2-4-6-5-3-1;/h4*1-5H;

InChI key

CRHIAMBJMSSNNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

231.8 °F - closed cup

Flash Point(C)

111 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuji Ohhira et al.
Journal of applied toxicology : JAT, 23(1), 31-35 (2003-01-09)
The metabolism of tetraphenyltin in rat liver and kidney has been examined. Tetraphenyltin and its metabolites in the tissue were determined periodically for 96 h after a single oral dose of 55.4 mg kg(-1) of tetraphenyltin by gas chromatography. The
A S Chawla et al.
Biomaterials, medical devices, and artificial organs, 13(3-4), 153-162 (1985-01-01)
Lactic acid was polymerized using tetraphenyl tin as a catalyst. The molecular weight of the resultant poly(lactic acid) varied between 0.89 X 10(6) and 2.94 X 10(6) depending upon the concentration of the catalyst used. In-vivo degradation of the poly(lactic
Mai Nozaki et al.
Journal of chromatography. A, 1622, 461173-461173 (2020-05-27)
Poly(diphenylacetylene) having optically active anilide pendants (poly-1) were synthesized by the condensation reaction of an optically active carboxylic acid with a key precursor polymer containing amino (-NH2) groups, which was prepared by the polymerization of a phthalimide-protected diphenylacetylene monomer using
Claudia Coughlan et al.
Chemical reviews, 117(9), 5865-6109 (2017-04-11)
This review captures the synthesis, assembly, properties, and applications of copper chalcogenide NCs, which have achieved significant research interest in the last decade due to their compositional and structural versatility. The outstanding functional properties of these materials stems from the
Manuel Gadogbe et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 54(9), 884-890 (2019-05-07)
Exposure to tin in the general US population is near ubiquitous, as determined using urinary tin levels measured by inductively coupled plasma mass spectrometry (ICP-MS). Urinary tin levels are associated with chronic health outcomes, such as diabetes; however, it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service