Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

P73803

Sigma-Aldrich

Pyrrolidine

99%

Synonym(s):

Tetrahydropyrrole, Tetramethyleneimine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9N
CAS Number:
Molecular Weight:
71.12
Beilstein:
102395
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.45 (vs air)

Quality Level

vapor pressure

128 mmHg ( 39 °C)
49 mmHg ( 20 °C)

Assay

99%

autoignition temp.

653 °F

expl. lim.

10.6 %

refractive index

n20/D 1.443 (lit.)

density

0.852 g/mL at 25 °C (lit.)

SMILES string

C1CCNC1

InChI

1S/C4H9N/c1-2-4-5-3-1/h5H,1-4H2

InChI key

RWRDLPDLKQPQOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyrrolidine is a heterocyclic building block used in organic synthesis and a scaffold for biologically active compounds.

Application

Pyrrolidine has been used for the synthesis of N-benzoyl pyrrolidine from benzaldehyde via oxidative amination. It may be used as a catalyst for the synthesis of N-sulfinyl aldimines from carbonyl compounds and sulfonamides.
Pyrrolidine can also be used to synthesize:
  • Taddol-pyrrolidine phosphoramidite, a ligand for rhodium-catalyzed [2+2+2] cycloaddition of pentenyl isocyanate and 4- ethynylanisole.
  • H,4 PyrrolidineQuin-BAM (′PBAM′), a selective catalyst for the aza-Henry addition of nitroalkanes to aryl aldimines.{88]
  • 1,2,3,3a,4,9-Hexahydropyrrolo[2,1-b]quinazoline by reacting with o-aminobenzaldehyde.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

37.4 °F - closed cup

Flash Point(C)

3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pyrrolidine in drug discovery: a versatile scaffold for novel biologically active compounds
Li Petri G, et al.
Topics in Current Chemistry, 379, 1-46 (2021)
Enantioselective Rhodium-Catalyzed [2+2+2] Cycloaddition of Pentenyl Isocyanate and 4-Ethynylanisole: Preparation and Use of Taddol-pyrrolidine Phosphoramidite
Oberg KM, et al.
Organic Syntheses, 91, 150-150 (2014)
Metal?Free One?Pot Oxidative Amination of Aromatic Aldehydes: Conversion of Benzaldehyde to N?Benzoyl Pyrrolidine.
Ekoue?Kovi K & Wolf C
Organic Syntheses, 1-7 (2010)
o-Aminobenzaldehyde, Redox-Neutral Aminal Formation and Synthesis of Deoxyvasicinone
Zhang C, et al.
Organic Syntheses, 89, 274-274 (2012)
Preparation of N-Sulfinyl Aldimines using Pyrrolidine as Catalyst via Iminium Ion Activation
Morales S, et al.
Organic Syntheses, 94, 346-346 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service