Skip to Content
MilliporeSigma
All Photos(2)

Documents

697478

Sigma-Aldrich

Tin(II) acetylacetonate

99.9% trace metals basis

Synonym(s):

Sn(acac)2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O4Sn
CAS Number:
Molecular Weight:
316.93
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

liquid

reaction suitability

core: tin

bp

110 °C/0.1 mmHg

SMILES string

CC(=O)\C=C(\C)O[SnH2]O\C(C)=C/C(C)=O

InChI

1S/2C5H8O2.Sn/c2*1-4(6)3-5(2)7;/h2*3,6H,1-2H3;/q;;+2/p-2/b2*4-3-;

InChI key

XDRPDDZWXGILRT-FDGPNNRMSA-L

Application

  • Thin film deposition by atomic layer deposition (ALD) 
  • Atomic layer etching.
  • For use in transistors and other integrated circuit components.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tobias Bubmann et al.
Polymers, 11(12) (2019-12-18)
The effect of different catalysts on reactive compatibilization of 50/50 polycarbonate (PC)/polymethylmethacrylate (PMMA) blends achieved via transesterification that occurs during compounding in a twin-screw extruder was investigated on a phenomenological (optical and mechanical properties), mesoscopic (phase morphology), and molecular level

Articles

The properties of many devices are limited by the intrinsic properties of the materials that compose them.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service