Key Documents
SYNA832440
Synthware inlet adapter with fritted disc
joint: ST/NS 24/40
Synonym(s):
Synthware A832440, glass inlet adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A832440
Related Categories
General description
With an extra coarse disc to prevent solid from being introduced into reaction system during liquid transfer.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)