Key Documents
SYNA322440
Synthware vacuum adapter
female joint: ST/NS 24/40
Synonym(s):
Synthware A322440, glass vacuum adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
female joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A322440
Related Categories
General description
Ideal for use as a vent or for the introduction of pressure/vacuum.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)