Key Documents
SYNA793524
Synthware bushing adapter
female joint: ST/NS 14/35, male joint: ST/NS 24/40
Synonym(s):
Synthware A793524, glass bushing adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Pricing and availability is not currently available.
Joint
female joint: ST/NS14/35
male joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A793524
Related Categories
General description
Bushing adapters allow for compact assemblies of dissimilar joints.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)