Key Documents
SYNA791419
Synthware bushing adapter
female joint: ST/NS 14/20, male joint: ST/NS 19/22
Synonym(s):
Synthware A791419, glass bushing adapter
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Joint
female joint: ST/NS14/20
male joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A791419
General description
Bushing adapters allow for compact assemblies of dissimilar joints.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)