Key Documents
Z530409
Sigma-Aldrich
Aldrich® short-path distillation head
Joints: ST/NS 29/32 (2), thermometer joint: ST/NS 10/30
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Joint
thermometer joint: ST/NS10/30
Joints: ST/NS29/32 (2)
immersion depth
75 mm
General description
Vigreux indents prevent bumping of fine or lightweight column packing into the condenser or fraction cutter when column is flooded.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)