Skip to Content
MilliporeSigma
All Photos(1)

Documents

S3757

Sigma-Aldrich

α-Solanine

from potato sprouts, ≥95%

Synonym(s):

alpha-Solanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H73NO15
CAS Number:
Molecular Weight:
868.06
Beilstein:
77607
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

biological source

potato sprouts

Assay

≥95%

storage temp.

−20°C

SMILES string

C[C@H]1CC[C@@H]2[C@@H](C)[C@H]3[C@H](C[C@H]4[C@@H]5CC=C6C[C@H](CC[C@]6(C)[C@H]5CC[C@]34C)O[C@@H]7O[C@H](CO)[C@H](O)[C@H](O[C@@H]8O[C@H](CO)[C@@H](O)[C@H](O)[C@H]8O)[C@H]7O[C@@H]9O[C@@H](C)[C@H](O)[C@@H](O)[C@H]9O)N2C1

InChI

1S/C45H73NO15/c1-19-6-9-27-20(2)31-28(46(27)16-19)15-26-24-8-7-22-14-23(10-12-44(22,4)25(24)11-13-45(26,31)5)57-43-40(61-41-37(54)35(52)32(49)21(3)56-41)39(34(51)30(18-48)59-43)60-42-38(55)36(53)33(50)29(17-47)58-42/h7,19-21,23-43,47-55H,6,8-18H2,1-5H3/t19-,20+,21-,23-,24+,25-,26-,27+,28-,29+,30+,31-,32-,33+,34-,35+,36-,37+,38+,39-,40+,41-,42-,43+,44-,45-/m0/s1

InChI key

ZGVSETXHNHBTRK-UDJLNJFBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Solanine is a steroidal glycoalkaloid mainly found in potatoes. It is a potential anti-carcinogenic agent against pancreatic cancer cells.

Application

α-Solanine has been used as a standard during its quantification in potatoes by HPLC.

Other Notes

A trisaccharide, consisting of glucose, galactose and rhamnose, linked to solanidine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kun-Hung Shen et al.
International journal of molecular sciences, 21(9) (2020-05-01)
Sinomenine is an alkaloid derived from Sinomeniumacutum. Recent studies have found that sinomenine can inhibit various cancers by inhibiting the proliferation, migration and invasion of tumors and inducing apoptosis. This study aims to investigate the effect and mechanism of sinomenine
Potatoes, tomatoes, and solanine toxicity (Solanum tuberosum L., Solanum lycopersicum L.).
Donald G Barceloux
Disease-a-month : DM, 55(6), 391-402 (2009-05-19)
Classification of potato tubers based on solanine toxicant using laser-induced light backscattering imaging.
Babazadeh S, et al.
Computers and Electronics in Agriculture, 129, 1-8 (2016)
Anupam Maurya et al.
Phytochemical analysis : PCA, 24(1), 87-92 (2012-07-13)
Solanum species are important ingredients of many traditional Indian medicines and thus the quality control of their herbal formulations is of paramount concern. To establish a simple and effective high-performance liquid chromatographic (HPLC) method to evaluate the quality of Solanum
Miriam Frida Karlsson et al.
Phytochemistry, 85, 72-81 (2012-11-06)
Guatemalan potato moth, Tecia solanivora, lay eggs in the soil nearby potato Solanum spp. and larvae feed on the tubers. We investigated the oviposition behaviour of T. solanivora females and the survival of larval offspring on healthy vs. stressed, i.e.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service