Skip to Content
MilliporeSigma
α-Solanine phyproof® Reference Substance

α-Solanine

α-Solanine phyproof® Reference Substance
Synonym(s):
alpha-Solanine
Empirical Formula (Hill Notation):
C45H73NO15
CAS Number:
Molecular Weight:
868.06
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Product Line
Purity
Format
Tissue
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing